Ethyl 4-methylvalerate 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 4-methylpentanoate
OR938286-25G
OR938286-5G
APOSOR938286-25GEA
206
EUR
APOSOR938286-25G
APOSOR938286-5G
Ethyl 4-methylvalerate 95%
Ethyl 4-methylvalerate
Formel:
C₈H₁₆O₂ Molecular Weight: 144,211 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042887 CAS-Nummer: 25415-67-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...