1-(Boc-amino)-2,3-butadiene 96%

Lieferant: Apollo Scientific

OR938527-250MG OR938527-500MG
APOSOR938527-250MGEA 219 EUR
APOSOR938527-250MG APOSOR938527-500MG
1-(Boc-amino)-2,3-butadiene 96%
1-(Boc-amino)-2,3-butadiene
Formel: C₉H₁₅NO₂
Molecular Weight: 169.221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18252670
CAS-Nummer: 92136-43-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR