3-Chloro-4-nitrobenzonitrile 98%
Lieferant: Apollo Scientific
OR938611-1G
OR938611-250MG
APOSOR938611-1GEA
122
EUR
APOSOR938611-1G
APOSOR938611-250MG
3-Chloro-4-nitrobenzonitrile 98%
3-Chloro-4-nitrobenzonitrile
Formel:
C₇H₃ClN₂O₂ Molecular Weight: 182,564 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02317167 CAS-Nummer: 34662-29-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...