(1S,4R,5R)-1-Isopropyl-4-methyl-bicyclo[3.1.0]hexan-3-one 95%
Lieferant: Apollo Scientific
OR938655-1G
OR938655-5G
APOSOR938655-1GEA
0
EUR
APOSOR938655-1G
APOSOR938655-5G
(1S,4R,5R)-1-Isopropyl-4-methyl-bicyclo[3.1.0]hexan-3-one 95%
(1S,4R,5R)-1-Isopropyl-4-methyl-bicyclo[3.1.0]hexan-3-one
Formel:
C₁₀H₁₆O Molecular Weight: 152,233 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001313 CAS-Nummer: 546-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...