4-Chloro-5-methoxy-1-indanone 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-5-methoxy-2,3-dihydro-1H-inden-1-one

OR938838-1G OR938838-5G
APOSOR938838-1GEA 126 EUR
APOSOR938838-1G APOSOR938838-5G
4-Chloro-5-methoxy-1-indanone 95%
4-Chloro-5-methoxy-1-indanone
Formel: C₁₀H₉ClO₂
Molecular Weight: 196,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09836228
CAS-Nummer: 944109-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR