3-(4-Hydroxy-3,5-dimethylphenyl)propanoic acid 95%
Lieferant: Apollo Scientific
OR938946-1G
OR938946-250MG
APOSOR938946-1GEA
632
EUR
APOSOR938946-1G
APOSOR938946-250MG
3-(4-Hydroxy-3,5-dimethylphenyl)propanoic acid 95%
3-(4-Hydroxy-3,5-dimethylphenyl)propanoic acid
Formel:
C₁₁H₁₄O₃ Molecular Weight: 194,227 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11188349 CAS-Nummer: 7733-58-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...