Quinoxalin-2-amine 95%

Lieferant: Apollo Scientific

OR938970-1G OR938970-250MG
APOSOR938970-1GEA 114 EUR
APOSOR938970-1G APOSOR938970-250MG
Quinoxalin-2-amine 95%
Quinoxalin-2-amine
Formel: C₈H₇N₃
Molecular Weight: 145,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00094018
CAS-Nummer: 5424-05-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR