2-Chloro-N-pyridin-2-ylacetamide 95%
Lieferant: Apollo Scientific
OR939010-1G
OR939010-5G
APOSOR939010-1GEA
261
EUR
APOSOR939010-1G
APOSOR939010-5G
2-Chloro-N-pyridin-2-ylacetamide 95%
2-Chloro-N-pyridin-2-ylacetamide
Formel:
C₇H₇ClN₂O Molecular Weight: 170,596 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023426 CAS-Nummer: 5221-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...