1-Bromo-4-phenylbutan-2-one 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR939073-1G
OR939073-250MG
OR939073-5G
APOSOR939073-1GEA
458
EUR
APOSOR939073-1G
APOSOR939073-250MG
APOSOR939073-5G
1-Bromo-4-phenylbutan-2-one 98%
1-Bromo-4-phenylbutan-2-one
Formel:
C₁₀H₁₁BrO Molecular Weight: 227,098 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11847199 CAS-Nummer: 31984-10-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...