2-Methyl-1H-benzo[d]imidazol-5-ol 98%

Lieferant: Apollo Scientific

OR939089-1G OR939089-250MG
APOSOR939089-1GEA 348 EUR
APOSOR939089-1G APOSOR939089-250MG
2-Methyl-1H-benzo[d]imidazol-5-ol 98%
2-Methyl-1H-benzo[d]imidazol-5-ol
Formel: C₈H₈N₂O
Molecular Weight: 148,162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12031291
CAS-Nummer: 41292-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR