2-((4-Methoxybenzyl)oxy)acetic acid 95%
Lieferant: Apollo Scientific
OR939102-5G
APOSOR939102-5GEA
55.1
EUR
APOSOR939102-5G
2-((4-Methoxybenzyl)oxy)acetic acid 95%
2-((4-Methoxybenzyl)oxy)acetic acid
Formel:
C₁₀H₁₂O₄ Molecular Weight: 196,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861881 CAS-Nummer: 88920-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...