4-(Prop-1-en-2-yl)phenol 95%

Lieferant: Apollo Scientific

OR939114-1G OR939114-250MG
APOSOR939114-1GEA 66.7 EUR
APOSOR939114-1G APOSOR939114-250MG
4-(Prop-1-en-2-yl)phenol 95%
4-(Prop-1-en-2-yl)phenol
Formel: C₉H₁₀O
Molecular Weight: 134,175 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00128236
CAS-Nummer: 4286-23-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR