2,3-Dimethylsuccinic acid 95%
Lieferant: Apollo Scientific
OR939184-1G
OR939184-5G
APOSOR939184-1GEA
203
EUR
APOSOR939184-1G
APOSOR939184-5G
2,3-Dimethylsuccinic acid 95%
2,3-Dimethylsuccinic acid
Formel:
C₆H₁₀O₄ Molecular Weight: 146,141 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002657 CAS-Nummer: 13545-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...