6-Chloro-3,4-dihydro-2H-isoquinolin-1-one 95%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-3,4-dihydroisoquinolin-1(2H)-one

OR939226-1G OR939226-5G
APOSOR939226-1GEA 72.5 EUR
APOSOR939226-1G APOSOR939226-5G
6-Chloro-3,4-dihydro-2H-isoquinolin-1-one 95%
6-Chloro-3,4-dihydro-2H-isoquinolin-1-one
Formel: C₉H₈ClNO
Molecular Weight: 181,619 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114866
CAS-Nummer: 22246-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR