tert-Butyl(4-iodobutoxy)dimethylsilane 97%

Lieferant: Apollo Scientific
Achtung

OR939273-25G OR939273-5G
APOSOR939273-25GEA 342 EUR
APOSOR939273-25G APOSOR939273-5G
tert-Butyl(4-iodobutoxy)dimethylsilane 97%
tert-Butyl(4-iodobutoxy)dimethylsilane
Formel: C₁₀H₂₃IOSi
Molecular Weight: 314,279 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01863695
CAS-Nummer: 92511-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR