4-Chloro-pentanoyl chloride 95%

Lieferant: Apollo Scientific

OR939296-1G OR939296-250MG OR939296-5G
APOSOR939296-1GEA 313 EUR
APOSOR939296-1G APOSOR939296-250MG APOSOR939296-5G
4-Chloro-pentanoyl chloride 95%
4-Chloro-pentanoyl chloride
Formel: C₅H₈Cl₂O
Molecular Weight: 155.022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08445304
CAS-Nummer: 63480-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR