Ethyl 3-cyano-2-oxopropanoate 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR939384-1G
OR939384-250MG
APOSOR939384-1GEA
0
EUR
APOSOR939384-1G
APOSOR939384-250MG
Ethyl 3-cyano-2-oxopropanoate 95%
Ethyl 3-cyano-2-oxopropanoate
Formel:
C₆H₇NO₃ Molecular Weight: 141,125 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03428327 CAS-Nummer: 56290-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...