(4,5-Dimethylthiazol-2-yl)methanol 95%
Lieferant: Apollo Scientific
OR939388-1G
OR939388-250MG
APOSOR939388-1GEA
139
EUR
APOSOR939388-1G
APOSOR939388-250MG
(4,5-Dimethylthiazol-2-yl)methanol 95%
(4,5-Dimethylthiazol-2-yl)methanol
Formel:
C₆H₉NOS Molecular Weight: 143,207 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12198452 CAS-Nummer: 99839-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...