3-Methyl-5-bromo-1,2,4-thiadiazole ≥98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-3-methyl-1,2,4-thiadiazole

OR939455-100MG OR939455-1G OR939455-250MG
APOSOR939455-100MGEA 21.9 EUR
APOSOR939455-100MG APOSOR939455-1G APOSOR939455-250MG
3-Methyl-5-bromo-1,2,4-thiadiazole ≥98%
3-Methyl-5-bromo-1,2,4-thiadiazole
Formel: C₃H₃BrN₂S
Molecular Weight: 179,038 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847500
CAS-Nummer: 54681-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR