(S)-4-Methyloxazolidin-2-one 95%
Lieferant: Apollo Scientific
OR939517-1G
APOSOR939517-1GEA
77
EUR
APOSOR939517-1G
(S)-4-Methyloxazolidin-2-one 95%
(S)-4-Methyloxazolidin-2-one
Formel:
C₄H₇NO₂ Molecular Weight: 101,104 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06656589 CAS-Nummer: 4042-35-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...