5-Nitrofurfuryl alcohol 96%

Lieferant: Apollo Scientific

Synonyme: (5-Nitrofuran-2-yl)methanol

OR939570-1G OR939570-250MG
APOSOR939570-1GEA 37.7 EUR
APOSOR939570-1G APOSOR939570-250MG
5-Nitrofurfuryl alcohol 96%
5-Nitrofurfuryl alcohol
Formel: C₅H₅NO₄
Molecular Weight: 143.098 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00272267
CAS-Nummer: 2493-04-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR