5-(Bromomethyl)isoxazole 95%

Lieferant: Apollo Scientific

OR939638-1G OR939638-250MG
APOSOR939638-1GEA 1250 EUR
APOSOR939638-1G APOSOR939638-250MG
5-(Bromomethyl)isoxazole 95%
5-(Bromomethyl)isoxazole
Formel: C₄H₄BrNO
Molecular Weight: 161,985 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669517
CAS-Nummer: 69735-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR