5-Nitro-1,2,3,4-tetrahydroisoquinoline 95%

Lieferant: Apollo Scientific

OR939642-1G OR939642-250MG
APOSOR939642-1GEA 348 EUR
APOSOR939642-1G APOSOR939642-250MG
5-Nitro-1,2,3,4-tetrahydroisoquinoline 95%
5-Nitro-1,2,3,4-tetrahydroisoquinoline
Formel: C₉H₁₀N₂O₂
Molecular Weight: 178.188 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06804518
CAS-Nummer: 41959-45-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR