2,3-Dihydro-1,4-isoquinolinedione 95%
Lieferant: Apollo Scientific
Synonyme:
2,3-Dihydroisoquinoline-1,4-dione
OR940009-1G
OR940009-250MG
APOSOR940009-1GEA
184
EUR
APOSOR940009-1G
APOSOR940009-250MG
2,3-Dihydro-1,4-isoquinolinedione 95%
2,3-Dihydro-1,4-isoquinolinedione
Formel:
C₉H₇NO₂ Molecular Weight: 161,157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13179340 CAS-Nummer: 31053-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...