6-Chloroimidazo[1,2-b]pyridazine-3-carbonitrile 95%
Lieferant: Apollo Scientific
OR940590-1G
OR940590-500MG
OR940590-250MG
APOSOR940590-1GEA
313
EUR
APOSOR940590-1G
APOSOR940590-500MG
APOSOR940590-250MG
6-Chloroimidazo[1,2-b]pyridazine-3-carbonitrile 95%
6-Chloroimidazo[1,2-b]pyridazine-3-carbonitrile
Formel:
C₇H₃ClN₄ Molecular Weight: 178,579 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11044763 CAS-Nummer: 123531-54-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...