2-Chloro-4,5-dimethyl-1,3-thiazole 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-4,5-dimethylthiazole

OR940618-1G OR940618-5G
APOSOR940618-1GEA 284 EUR
APOSOR940618-1G APOSOR940618-5G
2-Chloro-4,5-dimethyl-1,3-thiazole 95%
2-Chloro-4,5-dimethyl-1,3-thiazole
Formel: C₅H₆ClNS
Molecular Weight: 147.626 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17249933
CAS-Nummer: 56355-41-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR