Methyl 2-[3-(aminomethyl)phenyl]acetate hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-(3-(aminomethyl)phenyl)acetate hydrochloride

OR940759-1G OR940759-250MG
APOSOR940759-1GEA 763 EUR
APOSOR940759-1G APOSOR940759-250MG
Methyl 2-[3-(aminomethyl)phenyl]acetate hydrochloride 95%
Methyl 2-[3-(aminomethyl)phenyl]acetate hydrochloride
Formel: C₁₀H₁₃NO₂·ClH
Molecular Weight: 215.677 g/mol
MDL: MFCD09953145
CAS-Nummer: 197792-60-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR