3-(Cyclopropylmethoxy)-4-hydroxybenzaldehyde 95%
Lieferant: Apollo Scientific
OR940968-1G
OR940968-250MG
APOSOR940968-1GEA
148
EUR
APOSOR940968-1G
APOSOR940968-250MG
3-(Cyclopropylmethoxy)-4-hydroxybenzaldehyde 95%
3-(Cyclopropylmethoxy)-4-hydroxybenzaldehyde
Formel:
C₁₁H₁₂O₃ Molecular Weight: 192,211 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16999806 CAS-Nummer: 25934-52-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...