N-Methyl-2-nitrobenzylamine 97%

Lieferant: Apollo Scientific

Synonyme: N-Methyl-1-(2-nitrophenyl)methanamine

OR941225-1G OR941225-5G
APOSOR941225-1GEA 85.7 EUR
APOSOR941225-1G APOSOR941225-5G
N-Methyl-2-nitrobenzylamine 97%
N-Methyl-2-nitrobenzylamine
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166.177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039891
CAS-Nummer: 56222-08-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR