N,5-Dimethylpyridin-2-amine 95%

Lieferant: Apollo Scientific

OR941253-1G OR941253-250MG
APOSOR941253-1GEA 74.1 EUR
APOSOR941253-1G APOSOR941253-250MG
N,5-Dimethylpyridin-2-amine 95%
N,5-Dimethylpyridin-2-amine
Formel: C₇H₁₀N₂
Molecular Weight: 122,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06411230
CAS-Nummer: 45715-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR