(1-Isocyanatoethyl)benzene 95%

Lieferant: Apollo Scientific

OR941358-1G OR941358-250MG OR941358-500MG
APOSOR941358-1GEA 574 EUR
APOSOR941358-1G APOSOR941358-250MG APOSOR941358-500MG
(1-Isocyanatoethyl)benzene 95%
(1-Isocyanatoethyl)benzene

May form small amount of sediment

Formel: C₉H₉NO
Molecular Weight: 147.174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739387
CAS-Nummer: 1837-73-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR