5-Chloro-2-dimethylamino-phenol 95%

Lieferant: Apollo Scientific

OR941422-1G OR941422-250MG
APOSOR941422-1GEA 574 EUR
APOSOR941422-1G APOSOR941422-250MG
5-Chloro-2-dimethylamino-phenol 95%
5-Chloro-2-dimethylamino-phenol
Formel: C₈H₁₀ClNO
Molecular Weight: 171,624 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16998110
CAS-Nummer: 57649-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR