6-Bromo-2-methylpyridin-3-ol 97%

Lieferant: Apollo Scientific

Synonyme: 6-Bromo-3-hydoxy-2-methylpyridine

OR941712-1G OR941712-250MG
APOSOR941712-1GEA 159 EUR
APOSOR941712-1G APOSOR941712-250MG
6-Bromo-2-methylpyridin-3-ol 97%
6-Bromo-2-methylpyridin-3-ol
Formel: C₆H₆BrNO
Molecular Weight: 188.022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044277
CAS-Nummer: 118399-86-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR