6-Aminohexanal, N-BOC protected 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl N-(6-oxohexyl)carbamate

OR941901-250MG OR941901-1G
APOSOR941901-250MGEA 582 EUR
APOSOR941901-250MG APOSOR941901-1G
6-Aminohexanal, N-BOC protected 95%
6-Aminohexanal, N-BOC protected
Formel: C₁₁H₂₁NO₃
Molecular Weight: 215,289 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD12031715
CAS-Nummer: 80860-42-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR