Indan-1,2-dione-2-oxime 95%

Lieferant: Apollo Scientific

OR941928-10G OR941928-1G OR941928-5G
APOSOR941928-10GEA 451 EUR
APOSOR941928-10G APOSOR941928-1G APOSOR941928-5G
Indan-1,2-dione-2-oxime 95%
Indan-1,2-dione-2-oxime
Formel: C₉H₇NO₂
Molecular Weight: 161.157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00463407
CAS-Nummer: 15028-10-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR