2-Isopropoxybenzonitrile 95%

Lieferant: Apollo Scientific

OR942169-1G OR942169-250MG
APOSOR942169-1GEA 487 EUR
APOSOR942169-1G APOSOR942169-250MG
2-Isopropoxybenzonitrile 95%
2-Isopropoxybenzonitrile
Formel: C₁₀H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD02257394
CAS-Nummer: 90921-35-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR