3-Hydroxy-4-methyl-benzaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 3-Hydroxy-4-methylbenzaldehyde

OR942429-1G
APOSOR942429-1GEA 92.8 EUR
APOSOR942429-1G
3-Hydroxy-4-methyl-benzaldehyde 95%
3-Hydroxy-4-methyl-benzaldehyde
Formel: C₈H₈O₂
Molecular Weight: 136,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01632496
CAS-Nummer: 57295-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR