4-Oxo-1,4-dihydro-quinoline-8-carbonitrile 95%

Lieferant: Apollo Scientific

OR942656-1G OR942656-250MG
APOSOR942656-1GEA 377 EUR
APOSOR942656-1G APOSOR942656-250MG
4-Oxo-1,4-dihydro-quinoline-8-carbonitrile 95%
4-Oxo-1,4-dihydro-quinoline-8-carbonitrile
Formel: C₁₀H₆N₂O
Molecular Weight: 170.167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26131872
CAS-Nummer: 127285-55-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR