(S)-1-(5-Methylfuran-2-yl)propan-1-amine 95%

Lieferant: Apollo Scientific

OR942763-1G OR942763-250MG
APOSOR942763-1GEA 308 EUR
APOSOR942763-1G APOSOR942763-250MG
(S)-1-(5-Methylfuran-2-yl)propan-1-amine 95%
(S)-1-(5-Methylfuran-2-yl)propan-1-amine
Formel: C₈H₁₃NO
Molecular Weight: 139.195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11973975
CAS-Nummer: 473732-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR