6-Chloro-1-methylpyridin-2(1H)-one 95%

Lieferant: Apollo Scientific

OR943025-1G OR943025-250MG
APOSOR943025-1GEA 632 EUR
APOSOR943025-1G APOSOR943025-250MG
6-Chloro-1-methylpyridin-2(1H)-one 95%
6-Chloro-1-methylpyridin-2(1H)-one
Formel: C₆H₆ClNO
Molecular Weight: 143.571 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00955763
CAS-Nummer: 17228-63-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR