1-(2-Amino-4-methylphenyl)ethanone 95%

Lieferant: Apollo Scientific

OR943044-1G OR943044-5G
APOSOR943044-1GEA 192 EUR
APOSOR943044-1G APOSOR943044-5G
1-(2-Amino-4-methylphenyl)ethanone 95%
1-(2-Amino-4-methylphenyl)ethanone
Formel: C₉H₁₁NO
Molecular Weight: 149,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19204667
CAS-Nummer: 122710-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR