Dimethallylamine 95%

Lieferant: Apollo Scientific

Synonyme: Bis(2-methylallyl)amine

OR943096-10G OR943096-5G
APOSOR943096-10GEA 330 EUR
APOSOR943096-10G APOSOR943096-5G
Dimethallylamine 95%
Dimethallylamine
Formel: C₈H₁₅N
Molecular Weight: 125,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00026058
CAS-Nummer: 35000-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR