3-Nitro-DL-phenylalanine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-3-(3-nitrophenyl)propanoic acid

OR943164-1G OR943164-5G
APOSOR943164-1GEA 56.7 EUR
APOSOR943164-1G APOSOR943164-5G
3-Nitro-DL-phenylalanine 97%
3-Nitro-DL-phenylalanine
Formel: C₉H₁₀N₂O₄
Molecular Weight: 210,187 g/mol
Schmelzpunkt: 230…240 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00069003
CAS-Nummer: 22888-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR