7-Chlorobenzo[d]isoxazol-3-amine 95%

Lieferant: Apollo Scientific

OR943186-1G OR943186-250MG
APOSOR943186-1GEA 734 EUR
APOSOR943186-1G APOSOR943186-250MG
7-Chlorobenzo[d]isoxazol-3-amine 95%
7-Chlorobenzo[d]isoxazol-3-amine
Formel: C₇H₅ClN₂O
Molecular Weight: 168,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845367
CAS-Nummer: 927413-64-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR