2-Oxopropyl benzoate 95%

Lieferant: Apollo Scientific

OR943264-1G OR943264-5G
APOSOR943264-1GEA 82.8 EUR
APOSOR943264-1G APOSOR943264-5G
2-Oxopropyl benzoate 95%
2-Oxopropyl benzoate
Formel: C₁₀H₁₀O₃
Molecular Weight: 178,185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01660185
CAS-Nummer: 6656-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR