4-Hydroxy-3-methyl-5h-furan-2-one 95%

Lieferant: Apollo Scientific

Synonyme: 4-Hydroxy-3-methylfuran-2(5H)-one

OR943277-1G OR943277-5G
APOSOR943277-1GEA 85.7 EUR
APOSOR943277-1G APOSOR943277-5G
4-Hydroxy-3-methyl-5h-furan-2-one 95%
4-Hydroxy-3-methyl-5h-furan-2-one
Formel: C₅H₆O₃
Molecular Weight: 114,099 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01860143
CAS-Nummer: 516-09-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR