Allyl-prop-2-ynyl-amine hydrochloride 95%
Lieferant: Apollo Scientific
OR943284-1G
OR943284-5G
APOSOR943284-1GEA
270
EUR
APOSOR943284-1G
APOSOR943284-5G
Allyl-prop-2-ynyl-amine hydrochloride 95%
Allyl-prop-2-ynyl-amine hydrochloride
Formel:
C₆H₉N·ClH Molecular Weight: 131,603 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03596896 CAS-Nummer: 1158414-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...