6-Methoxy-2-phenyl-tetralone

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-methoxy-2-phenyl-3,4-dihydro-2H-naphthalen-1-one

OR943451-5G
APOSOR943451-5GEA 185 EUR
APOSOR943451-5G
6-Methoxy-2-phenyl-tetralone
6-Methoxy-2-phenyl-tetralone
Formel: C₁₇H₁₆O₂
Molecular Weight: 252,308 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10698841
CAS-Nummer: 1769-84-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR