2-(1-Methyl-1H-pyrrol-2-yl)ethanamine 95%

Lieferant: Apollo Scientific

OR943470-1G OR943470-250MG
APOSOR943470-1GEA 734 EUR
APOSOR943470-1G APOSOR943470-250MG
2-(1-Methyl-1H-pyrrol-2-yl)ethanamine 95%
2-(1-Methyl-1H-pyrrol-2-yl)ethanamine
Formel: C₇H₁₂N₂
Molecular Weight: 124,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009753
CAS-Nummer: 83732-75-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR