(5-Chloro-2-methylphenyl)methanol 95%
Lieferant: Apollo Scientific
OR943490-1G
OR943490-250MG
APOSOR943490-1GEA
84.1
EUR
APOSOR943490-1G
APOSOR943490-250MG
(5-Chloro-2-methylphenyl)methanol 95%
(5-Chloro-2-methylphenyl)methanol
Formel:
C₈H₉ClO Molecular Weight: 156,609 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11111046 CAS-Nummer: 58966-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...